Proteintech
Manufacturer Code:666231IG150UL
Catalog # 50-173-7041
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1E1A4 |
Immunogen | CHST4 Fusion Protein Ag3987 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: carbohydrate (N-acetylglucosamine 6-O) sulfotransferase 4; Carbohydrate sulfotransferase 4; CHST4 GlcNAc6ST 2 Gn6st 2 GST 3 HEC GlcNAc6ST LSST; galactose/N-acetylglucosamine/N-acetylgalactosamine 6-O-sulfotransferase 3; Galactose/N-acetylglucosamine/N-acetylglucosamine 6-O-sulfotransferase 3; GlcNAc6ST-2; gn6st-2; GST-3; HEC-GlcNAc6ST; High endothelial cells N-acetylglucosamine 6-O-sulfotransferase; L-selectin ligand sulfotransferase; LSST; N-acetylglucosamine 6-O-sulfotransferase 2
Gene Aliases: CHST4; GlcNAc6ST2; GST3; HECGLCNAC6ST; LSST
UniProt ID: (Human) Q8NCG5
Entrez Gene ID: (Human) 10164
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.