Invitrogen
Manufacturer Code:PA541119
Catalog # PIPA541119
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human CHST4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: carbohydrate (chondroitin 6/keratan) sulfotransferase 4; carbohydrate (N-acetylglucosamine 6-O) sulfotransferase 4; Carbohydrate (N-acetylglucosamine 6-O) sulfotransferase 4 GST3 LSST GlcNAc6ST2 HECGLCNAC6ST; Carbohydrate sulfotransferase 4; galactose/N-acetylglucosamine/N-acetylgalactosamine 6-O-sulfotransferase 3; Galactose/N-acetylglucosamine/N-acetylglucosamine 6-O-sulfotransferase 3; GlcNAc6ST-2; gn6st-2; GST-3; HEC-GlcNAc6ST; high endothelial cell GlcNAC-6-sulphotransferase; High endothelial cells N-acetylglucosamine 6-O-sulfotransferase; L-selectin ligand sulfotransferase; LSST; N-acetylglucosamine 6-O-sulfotransferase 2
Gene Aliases: CHST4; GlcNAc6ST2; Gn6st-2; GST-3; GST3; HEC-GlcNAc6ST; HECGLCNAC6ST; LSST; sulfo6
UniProt ID: (Human) Q8NCG5, (Mouse) Q9R1I1
Entrez Gene ID: (Rabbit) 100345750, (Dog) 489722, (Bovine) 539063, (Horse) 100068136, (Human) 10164, (Pig) 106510480, (Guinea pig) 100719794, (Rat) 307838, (Mouse) 26887
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.