Invitrogen
Manufacturer Code:PA569407
Catalog # PIPA569407
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human CHST2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C6ST GST-2 GST2 Gn6ST-1 glcNAc6ST-1 N-acetylglucosamine 6-O-sulfotransferase 1 carbohydrate (chondroitin 6/keratan) sulfotransferase 2 carbohydrate sulfotransferase 2 galactose/N-acetylglucosamine/N-acetylglucosamine 6-O-sulfotransferase 2; carbohydrate (chondroitin 6/keratan) sulfotransferase 2; carbohydrate (N-acetylglucosamine-6-O) sulfotransferase 2; Carbohydrate sulfotransferase 2; epididymis secretory protein Li 75; Galactose/N-acetylglucosamine/N-acetylglucosamine 6-O-sulfotransferase 2; GlcNAc6ST-1; gn6st-1; GST-2; N-acetylglucosamine 6-O-sulfotransferase 1; N-acetylglucosamine-6-O-sulfotransferase
Gene Aliases: AI428561; AW121776; C130041E03Rik; C6ST; CHST2; Chts2; GlcNAc6ST; glcNAc6ST-1; GN6ST; Gn6ST-1; GST-2; GST2; HEL-S-75
UniProt ID: (Human) Q9Y4C5, (Mouse) Q80WV3
Entrez Gene ID: (Dog) 485701, (Horse) 100147045, (Human) 9435, (Bovine) 615978, (Rabbit) 100355215, (Mouse) 54371, (Guinea pig) 100714052, (Rat) 367145
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.