Invitrogen
Manufacturer Code:PA564950
Catalog # PIPA564950
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit Human CHST2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C6ST GST-2 GST2 Gn6ST-1 glcNAc6ST-1 N-acetylglucosamine 6-O-sulfotransferase 1 carbohydrate (chondroitin 6/keratan) sulfotransferase 2 carbohydrate sulfotransferase 2 galactose/N-acetylglucosamine/N-acetylglucosamine 6-O-sulfotransferase 2; carbohydrate (chondroitin 6/keratan) sulfotransferase 2; carbohydrate (N-acetylglucosamine-6-O) sulfotransferase 2; Carbohydrate sulfotransferase 2; epididymis secretory protein Li 75; Galactose/N-acetylglucosamine/N-acetylglucosamine 6-O-sulfotransferase 2; GlcNAc6ST-1; GST-2; N-acetylglucosamine 6-O-sulfotransferase 1
Gene Aliases: C6ST; CHST2; glcNAc6ST-1; GN6ST; Gn6ST-1; GST-2; GST2; HEL-S-75
UniProt ID: (Human) Q9Y4C5
Entrez Gene ID: (Human) 9435
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.