Invitrogen
Manufacturer Code:PA597171
Catalog # PIPA597171
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 452-561 of human CHST15. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: B cell RAG associated protein; B cell RAG associated protein (GALNAC4S-6ST); B-cell RAG-associated gene protein; Carbohydrate sulfotransferase 15; GalNAc4S-6ST; hBRAG; N-acetylgalactosamine 4-sulfate 6-O-sulfotransferase
Gene Aliases: BRAG; CHST15; GALNAC4S-6ST; GALNAC4S6ST; KIAA0598
UniProt ID: (Human) Q7LFX5, (Rat) Q8CHI9
Entrez Gene ID: (Human) 51363, (Rat) 286974
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.