Invitrogen
Manufacturer Code:PA553609
Catalog # PIPA553609
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human CHST15 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: B cell RAG associated protein (GALNAC4S-6ST); B-cell RAG-associated gene protein; BRAG GALNAC4S-6ST RP11-47G11.1 B cell RAG associated protein (GALNAC4S-6ST) B-cell RAG-associated gene protein N-acetylgalactosamine 4-sulfate 6-O-sulfotransferase carbohydrate sulfotransferase 15 hBRAG; Carbohydrate sulfotransferase 15; GalNAc4S-6ST; hBRAG; N-acetylgalactosamine 4-sulfate 6-O-sulfotransferase
Gene Aliases: BRAG; CHST15; GALNAC4S-6ST; GALNAC4S6ST; KIAA0598
UniProt ID: (Human) Q7LFX5
Entrez Gene ID: (Human) 51363
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.