Invitrogen
Manufacturer Code:PA542989
Catalog # PIPA542989
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human GALNAC4S-6ST |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: B cell RAG associated protein; B cell RAG associated protein (GALNAC4S-6ST); B-cell RAG-associated gene protein; Carbohydrate (N-acetylgalactosamine 4-sulfate 6-O) sulfotransferase 15 BRAG DKFZp781H1369 KIAA0598 MGC34346 RP11-47G11.1 GALNAC4S-6ST; Carbohydrate sulfotransferase 15; GalNAc 4-sulfate 6-O-sulfotransferase; GalNAc4S-6ST; hBRAG; N-acetylgalactosamine 4-sulfate 6-O-sulfotransferase
Gene Aliases: 4631426J05Rik; BRAG; CHST15; GALNAC4S-6ST; GALNAC4S6ST; GalNAcS-6ST; KIAA0598; MAd5; mKIAA0598
UniProt ID: (Human) Q7LFX5, (Rat) Q8CHI9, (Mouse) Q91XQ5
Entrez Gene ID: (Dog) 486928, (Horse) 100057594, (Human) 51363, (Rabbit) 100343176, (Guinea pig) 100713991, (Rat) 286974, (Mouse) 77590
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.