Proteintech
Manufacturer Code:177491AP150UL
Catalog # 50-173-1763
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CHST14 Fusion Protein Ag12014 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: carbohydrate (dermatan 4) sulfotransferase 14; carbohydrate (N-acetylgalactosamine 4-0) sulfotransferase 14; Carbohydrate sulfotransferase 14; CHST14 D4ST 1 D4ST1 Dermatan 4 sulfotransferase 1 HD4ST hD4ST1 HNK1ST; D4ST-1; dermatan 4 sulfotransferase 1; Dermatan 4-sulfotransferase 1; dermatan-4-sulfotransferase-1
Gene Aliases: 2600016L03Rik; ATCS; CHST14; D4ST-1; D4ST1; EDSMC1; HNK1ST; UNQ1925/PRO4400
UniProt ID: (Human) Q8NCH0, (Mouse) Q80V53
Entrez Gene ID: (Human) 113189, (Mouse) 72136, (Rat) 691394
Molecular Function:
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.