Proteintech
Manufacturer Code:153411AP150UL
Catalog # 50-173-0161
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CHST12 Fusion Protein Ag7544 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C4S 2 C4ST 2 C4ST2 CHST12 Sulfotransferase Hlo; C4ST-2; Carbohydrate sulfotransferase 12; Chondroitin 4-O-sulfotransferase 2; Chondroitin 4-sulfotransferase 2; Sulfotransferase Hlo
Gene Aliases: AI595374; C4S-2; C4ST-2; C4ST2; CHST12; UNQ500/PRO1017
UniProt ID: (Human) Q9NRB3, (Mouse) Q99LL3
Entrez Gene ID: (Human) 55501, (Mouse) 59031, (Rat) 304322
Molecular Function:
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.