Invitrogen
Manufacturer Code:PA553002
Catalog # PIPA553002
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human CHST10 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Carbohydrate sulfotransferase 10; carbohydrate sulfotransferase 10 CHST10; HNK-1 sulfotransferase; HNK-1ST; HNK1ST; huHNK-1ST; raHNK-1ST; sul-T
Gene Aliases: AI507003; AU041319; CHST10; HNK-1ST; HNK1ST; ST
UniProt ID: (Human) O43529, (Rat) O54702, (Mouse) Q6PGK7
Entrez Gene ID: (Human) 9486, (Rat) 140568, (Mouse) 98388
Molecular Function: transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.