Invitrogen
Manufacturer Code:PA5106630
Catalog # PIPA5106630
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human CHST10 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AI507003 AU041319 Carbohydrate sulfotransferase 10 Chst10 HNK-1 sulfotransferase HNK1ST HNK-1ST huHNK-1ST raHNK-1ST ST sul-T; Carbohydrate sulfotransferase 10; HNK-1 sulfotransferase; HNK-1ST; HNK1ST; huHNK-1ST; raHNK-1ST; sul-T
Gene Aliases: AI507003; AU041319; CHST10; HNK-1ST; HNK1ST; ST
UniProt ID: (Human) O43529, (Rat) O54702, (Mouse) Q6PGK7
Entrez Gene ID: (Human) 9486, (Rat) 140568, (Mouse) 98388
Molecular Function: transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.