Novus Biologicals
Manufacturer Code:NBP2826870.1ML
Catalog # NB030616
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the N terminal region of human CHSS2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Chondroitin glucuronyltransferase 2; Chondroitin glucuronyltransferase 2 chondroitin polymerizing factor chondroitin sulfate synthase 2 Chondroitin-polymerizing factor ChPF CHSY2 CSS2FLJ22678 EC 2.4.1.175 EC 2.4.1.226 Glucuronosyl-N-acetylgalactosaminyl-proteoglycan4-beta-N-acetylgalactosaminyltransferase II N-acetylgalactosaminyl-proteoglycan 3-beta-glucuronosyltransferase II N-acetylgalactosaminyltransferase 2; Chondroitin sulfate synthase 2; Chondroitin-polymerizing factor; ChPF; Glucuronosyl-N-acetylgalactosaminyl-proteoglycan 4-beta-N-acetylgalactosaminyltransferase II; N-acetylgalactosaminyl-proteoglycan 3-beta-glucuronosyltransferase II; N-acetylgalactosaminyltransferase 2
Gene Aliases: CHPF; CHSY2; CSS2; UNQ651/PRO1281
UniProt ID: (Human) Q8IZ52
Entrez Gene ID: (Human) 79586
Molecular Function:
glycosyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.