Abnova
Manufacturer Code:H00001145M02
Catalog # 89-123-145
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1H5 |
Immunogen | CHRNE (NP_000071 21 a.a. ~ 130 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a partial recombinant CHRNE.
For Research Use Only
Protein Aliases: Acetylcholine receptor subunit epsilon; acetylcholine receptor, nicotinic, epsilon (muscle); AchR epsilon subunit; ACHRE; cholinergic receptor, nicotinic epsilon; cholinergic receptor, nicotinic, epsilon (muscle); cholinergic receptor, nicotinic, epsilon polypeptide; CMS1D; CMS1E; CMS2A; FCCMS; SCCMS
Gene Aliases: ACHRE; CHRNE; CMS1D; CMS1E; CMS2A; CMS4A; CMS4B; CMS4C; FCCMS; SCCMS
UniProt ID: (Human) Q04844
Entrez Gene ID: (Human) 1145
Molecular Function:
GABA receptor
acetylcholine receptor
ion channel
ligand-gated ion channel
receptor
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.