Invitrogen
Manufacturer Code:PA576134
Catalog # PIPA576134
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein of human CHRNE. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: acetylcholine receptor epsilon; Acetylcholine receptor subunit epsilon; Acetylcholine receptor subunit epsilon CHRNE ACHRE; acetylcholine receptor, nicotinic, epsilon (muscle); AchR epsilon subunit; cholinergic receptor, nicotinic epsilon; cholinergic receptor, nicotinic, epsilon (muscle); cholinergic receptor, nicotinic, epsilon polypeptide; epsilon-subunit; gene for acetylcholine receptor epsilon subunit; nicotinic acetylcholine receptor epsilon subunit
Gene Aliases: ACHRE; AChrepsilon; Acre; CHRNE; CMS1D; CMS1E; CMS2A; CMS4A; CMS4B; CMS4C; FCCMS; nAChRE; SCCMS
UniProt ID: (Human) Q04844, (Mouse) P20782, (Rat) P09660
Entrez Gene ID: (Human) 1145, (Mouse) 11448, (Rat) 29422
Molecular Function:
GABA receptor
acetylcholine receptor
ion channel
ligand-gated ion channel
receptor
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.