Invitrogen
Manufacturer Code:PA546828
Catalog # PIPA546828
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human CHRND |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: acetylcholine receptor delta subunit; Acetylcholine receptor subunit delta; acetylcholine receptor, nicotinic, delta (muscle); Cholinergic receptor nicotinic delta (muscle) ACHRD CMS2A FCCMS SCCMS; cholinergic receptor, nicotinic delta; cholinergic receptor, nicotinic, delta (muscle); cholinergic receptor, nicotinic, delta polypeptide; nicotinic acetylcholine receptor delta polypeptide; sofa potato; sop
Gene Aliases: ACHRD; Acrd; CHRND; CMS2A; CMS3A; CMS3B; CMS3C; FCCMS; SCCMS; wu:fa01e01; zgc:110469
UniProt ID: (Bovine) P04759, (Human) Q07001, (Rat) P25110
Entrez Gene ID: (Pig) 100522042, (Dog) 486162, (Bovine) 281687, (Rabbit) 100352617, (Human) 1144, (Guinea pig) 100733810, (Rat) 54240, (Zebrafish) 404273
Molecular Function: GABA receptor acetylcholine receptor ion channel ligand-gated ion channel receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.