Invitrogen
Manufacturer Code:PA528829
Catalog # PIPA528829
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to a region within amino acids 453 and 517 of Human CHRND |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acetylcholine receptor subunit delta; acetylcholine receptor, nicotinic, delta (muscle); ACHRD CMS2A FCCMS SCCMS acetylcholine receptor subunit delta acetylcholine receptor nicotinic delta (muscle) cholinergic receptor nicotinic delta polypeptide; cholinergic receptor, nicotinic delta; cholinergic receptor, nicotinic, delta (muscle); cholinergic receptor, nicotinic, delta polypeptide
Gene Aliases: ACHRD; CHRND; CMS2A; CMS3A; CMS3B; CMS3C; FCCMS; SCCMS
UniProt ID: (Human) Q07001
Entrez Gene ID: (Human) 1144
Molecular Function: GABA receptor acetylcholine receptor ion channel ligand-gated ion channel receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.