Invitrogen
Manufacturer Code:MA531695
Catalog # PIMA531695
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1H11F2 |
Immunogen | Purified recombit fragment of human CHRNB4 (AA: extra 22-236) expressed in E. coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: acetylcholine receptor, nicotinic, beta 4 (neuronal); cholinergic receptor, nicotinic beta 4; cholinergic receptor, nicotinic, beta 4 (neuronal); cholinergic receptor, nicotinic, beta polypeptide 4; Neuronal acetylcholine receptor subunit beta-4; neuronal nicotinic receptor beta 4 subunit
Gene Aliases: CHRNB4
UniProt ID: (Human) Q8WXC8
Entrez Gene ID: (Human) 1143
Molecular Function:
GABA receptor
acetylcholine receptor
ion channel
ligand-gated ion channel
receptor
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.