Invitrogen
Manufacturer Code:PA5103971
Catalog # PIPA5103971
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human CHRNB4 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: acetylcholine receptor Acetylcholine receptor beta 4 acetylcholine receptor nicotinic beta 4 (neuronal) Acrb4 Acrb-4 cholinergic receptor cholinergic receptor nicotinic beta 4 subunit cholinergic receptor nicotinic beta 4 cholinergic receptor nicotinic beta 4 (neuronal) cholinergic receptor nicotinic beta polypeptide 4 Chrnb4 nAChRB4 N-alpha 2 Neuronal acetylcholine receptor non-alpha-2 chain Neuronal acetylcholine receptor subunit beta-4 neuronal nicotinic receptor beta 4 subunit nicotinic acetylcholine receptor beta4 subunit; Acetylcholine receptor beta 4; acetylcholine receptor, nicotinic, beta 4 (neuronal); cholinergic receptor, nicotinic beta 4; cholinergic receptor, nicotinic, beta 4 (neuronal); cholinergic receptor, nicotinic, beta polypeptide 4; N-alpha 2; Neuronal acetylcholine receptor non-alpha-2 chain; Neuronal acetylcholine receptor subunit beta-4; neuronal nicotinic receptor beta 4 subunit; nicotinic acetylcholine receptor beta4 subunit
Gene Aliases: Acrb-4; Acrb4; CHRNB4
UniProt ID: (Human) Q8WXC8, (Mouse) Q8R493, (Rat) P12392
Entrez Gene ID: (Human) 1143, (Mouse) 108015, (Rat) 25103
Molecular Function:
GABA receptor
acetylcholine receptor
ion channel
ligand-gated ion channel
receptor
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.