Invitrogen
Manufacturer Code:PA595642
Catalog # PIPA595642
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Fusion protein from the cytoplasmic loop of the beta 2 subunit of rat nAChR |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: acetylcholine receptor beta 2 neural; cholinergic receptor nicotinic beta polypeptide 2; cholinergic receptor, nicotinic, beta polypeptide 2 (neuronal); cholinergic receptor, nicotinic, beta subunit 2; N-alpha 1; Neuronal acetylcholine receptor non-alpha-1 chain; Neuronal acetylcholine receptor subunit beta-2
Gene Aliases: [b]2-nAchR; Acrb-2; Acrb2; C030030P04Rik; Chrnb2
UniProt ID: (Rat) P12390, (Mouse) Q9ERK7
Entrez Gene ID: (Rat) 54239, (Mouse) 11444
Molecular Function: GABA receptor acetylcholine receptor ion channel ligand-gated ion channel receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.