Invitrogen
Manufacturer Code:PA529792
Catalog # PIPA529792
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 25 and 273 of Human Nicotinic Acetylcholine Receptor beta 2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: acetylcholine receptor beta 2 neural; EFNL3 nAChRB2 acetylcholine receptor nicotinic beta 2 (neuronal) cholinergic receptor nicotinic beta polypeptide 2 (neuronal) neuronal acetylcholine receptor subunit beta-2 neuronal nicotinic acetylcholine receptor beta 2; Neuronal acetylcholine receptor subunit beta-2
Gene Aliases: [b]2-nAchR; Acrb-2; Acrb2; C030030P04Rik; Chrnb2
UniProt ID: (Mouse) Q9ERK7
Entrez Gene ID: (Mouse) 11444
Molecular Function: GABA receptor acetylcholine receptor ion channel ligand-gated ion channel receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.