Invitrogen
Manufacturer Code:PA576463
Catalog # PIPA576463
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein of human CHRNA9. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: acetylcholine receptor alpha 9 subunit (nAChR); acetylcholine receptor, neuronal nicotinic, alpha-9 subunit; cholinergic receptor, nicotinic alpha 9; cholinergic receptor, nicotinic, alpha 9 (neuronal); cholinergic receptor, nicotinic, alpha polypeptide 9; NACHR alpha 9; NACHR alpha-9; neuronal acetylcholine receptor protein, alpha-9 subunit; Neuronal acetylcholine receptor subunit alpha-9; Neuronal acetylcholine receptor subunit alpha-9 Nicotinic acetylcholine receptor subunit alpha-9 NACHR alpha-9 CHRNA9 NACHRA9; nicotinic acetylcholine receptor subunit alpha 9; Nicotinic acetylcholine receptor subunit alpha-9
Gene Aliases: 2410015I05Rik; Acra9; CHRNA9; EG666827; Gm8311; HSA243342; NACHRA9
UniProt ID: (Human) Q9UGM1, (Rat) P43144
Entrez Gene ID: (Human) 55584, (Mouse) 231252, (Rat) 65024
Molecular Function: GABA receptor acetylcholine receptor ion channel ligand-gated ion channel receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.