Invitrogen
Manufacturer Code:PA546826
Catalog # PIPA546826
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human CHRNA9 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cholinergic receptor nicotinic alpha 9 (neuronal) NACHRA9 HSA243342; cholinergic receptor, nicotinic, alpha 9; cholinergic receptor, nicotinic, alpha 9 (neuronal); neuronal acetylcholine receptor subunit alpha-9
Gene Aliases: 2410015I05Rik; Acra9; chrna9; EG666827; Gm8311; si:dkey-274k22.9
Entrez Gene ID: (Rabbit) 100356862, (Bovine) 521876, (Dog) 488832, (Horse) 100054940, (Mouse) 231252, (Guinea pig) 100736216, (Zebrafish) 568807
Molecular Function:
GABA receptor
acetylcholine receptor
ion channel
ligand-gated ion channel
receptor
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.