Invitrogen
Manufacturer Code:PA5116644
Catalog # PIPA5116644
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 23-230 of human CHRNA7 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: a7 nicotinic acetylcholine receptor; a7 nicotinic acetylcholine receptor acetylcholine receptor acetylcholine receptor alpha 7 neural Acra7 alpha 7 neuronal nicotinic acetylcholine receptor alpha7 alpha-7 nicotinic cholinergic receptor subunit alpha7 nicotinic receptor alpha7-nAChR BTX bungarotoxin alpha C holinergic receptor nicotinic alpha polypeptide 7 (neuronal nicotinic acetycholine receptor alpha 7) (bungarotoxin alpha) C holinergic receptor nicotinic alpha polypeptide 7 (neuronal nicotinic acetycholine receptor alpha 7) (bungarotoxin alpha) cholinergic receptor cholinergic receptor nicotinic alpha 7 subunit cholinergic receptor nicotinic alpha 7 cholinergic receptor nicotinic alpha 7 (neuronal) cholinergic receptor nicotinic alpha polypeptide 7 CHRFAM7A CHRNA7 CHRNA7-2 CHRNA7-DR1 D-10 nAchR nAChR alpha7 NACHRA7 NARAD neuronal acetylcholine receptor protein alpha-7 chain neuronal acetylcholine receptor subunit alpha-7 neuronal nicotinic acetycholine receptor alpha 7 nicotinic receptor alpha 7 subunit; acetylcholine receptor alpha 7 neural; alpha 7 neuronal nicotinic acetylcholine receptor; alpha-7 nicotinic cholinergic receptor subunit; alpha7 nicotinic receptor; alpha7-nAChR; bungarotoxin alpha; C holinergic receptor nicotinic alpha polypeptide 7 (neuronal nicotinic acetycholine receptor alpha 7) (bungarotoxin alpha); C holinergic receptor, nicotinic, alpha polypeptide 7 (neuronal nicotinic acetycholine receptor alpha 7) (bungarotoxin alpha); cholinergic receptor, nicotinic alpha 7; cholinergic receptor, nicotinic, alpha 7 (neuronal); cholinergic receptor, nicotinic, alpha polypeptide 7; neuronal acetylcholine receptor protein, alpha-7 chain; Neuronal acetylcholine receptor subunit alpha-7; neuronal nicotinic acetycholine receptor alpha 7; nicotinic receptor alpha 7 subunit
Gene Aliases: Acra7; alpha7; BTX; CHRNA7; CHRNA7-2; nAchR; NACHRA7; NARAD
UniProt ID: (Human) P36544, (Rat) Q05941, (Mouse) P49582
Entrez Gene ID: (Human) 1139, (Rat) 25302, (Mouse) 11441
Molecular Function:
GABA receptor
acetylcholine receptor
ion channel
ligand-gated ion channel
receptor
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.