Proteintech
Manufacturer Code:135161AP150UL
Catalog # 50-172-8803
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CHRNA5 Fusion Protein Ag4447 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: acetylcholine receptor alpha 5; acetylcholine receptor, nicotinic, alpha 5 (neuronal); Cholinergic receptor, neuronal nicotinic, alpha polypeptide-5; cholinergic receptor, nicotinic alpha 5; cholinergic receptor, nicotinic, alpha 5 (neuronal); cholinergic receptor, nicotinic, alpha polypeptide 5; CHRNA5 LNCR2 NACHRA5; Neuronal acetylcholine receptor subunit alpha-5; neuronal nicotinic acetylcholine receptor, alpha5 subunit; nicotinic acetylcholine receptor alpha5 subunit
Gene Aliases: Acra-5; Acra5; CHRNA5; LNCR2; NACHRA5
UniProt ID: (Human) P30532, (Mouse) Q2MKA5
Entrez Gene ID: (Human) 1138, (Mouse) 110835, (Rat) 25102
Molecular Function: GABA receptor acetylcholine receptor ion channel ligand-gated ion channel receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.