Invitrogen
Manufacturer Code:MA537815
Catalog # PIMA537815
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC1822 |
Immunogen | A synthesized peptide derived from human ACHA4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: a4 nicotinic receptor; a4 nicotinic receptor acetylcholine receptor acetylcholine receptor alpha 4 neural acetylcholine receptor neuronal nicotinic alpha-4 subunit Acra4 Acra-4 alpha4 nAChR alpha4-nAChR BFNC cholinergic receptor cholinergic receptor nicotinic alpha 4 subunit cholinergic receptor nicotinic alpha 4 cholinergic receptor nicotinic alpha 4 (neuronal) cholinergic receptor nicotinic alpha 4 subunit cholinergic receptor nicotinic alpha polypeptide 4 CHRNA 4 CHRNA4 EBN EBN1 ENFL1 NACHR nAChR alpha 4 NACHRA4 NACRA4 NARAC Neuronal acetylcholine receptor subunit alpha-4 Neuronal nicotinic acetylcholine receptor alpha 4 subunit neuronal nicotinic acetylcholine receptor alpha-4 subunit; acetylcholine receptor alpha 4 neural; acetylcholine receptor, neuronal, nicotinic, alpha-4 subunit; alpha4 nAChR; alpha4-nAChR; cholinergic receptor, nicotinic alpha 4; cholinergic receptor, nicotinic, alpha 4 (neuronal); cholinergic receptor, nicotinic, alpha 4 subunit; cholinergic receptor, nicotinic, alpha polypeptide 4; Neuronal acetylcholine receptor subunit alpha-4; Neuronal nicotinic acetylcholine receptor alpha 4 subunit; neuronal nicotinic acetylcholine receptor alpha-4 subunit
Gene Aliases: Acra-4; Acra4; BFNC; CHRNA4; EBN; EBN1; ENFL1; NACHR; NACHRA4; NACRA4; NARAC
UniProt ID: (Human) P43681, (Mouse) O70174, (Rat) P09483
Entrez Gene ID: (Human) 1137, (Mouse) 11438, (Rat) 25590
Molecular Function:
GABA receptor
acetylcholine receptor
ion channel
ligand-gated ion channel
receptor
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.