Invitrogen
Manufacturer Code:PA572290
Catalog # PIPA572290
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 176-208 amino acids from human CHRNA4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: a4 nicotinic receptor; acetylcholine receptor alpha 4 neural; acetylcholine receptor, neuronal, nicotinic, alpha-4 subunit; alpha4 nAChR; alpha4-nAChR; cholinergic receptor, nicotinic alpha 4; cholinergic receptor, nicotinic, alpha 4 (neuronal); cholinergic receptor, nicotinic, alpha polypeptide 4; CHRNA4 cholinergic receptor nicotinic alpha 4 BFNC EBN EBN1 NACRA4cholinergic receptor nicotinic alpha 4 subunit cholinergic receptor nicotinic alpha polypeptide 4 neuronal acetylcholine receptor alpha 4 subunit neuronal nicotinic acetylcholine receptor alpha-4 subunit; Neuronal acetylcholine receptor subunit alpha-4; neuronal nicotinic acetylcholine receptor alpha-4 subunit
Gene Aliases: Acra-4; Acra4; BFNC; CHRNA4; EBN; EBN1; ENFL1; NACHR; NACHRA4; NACRA4
UniProt ID: (Human) P43681, (Mouse) O70174
Entrez Gene ID: (Human) 1137, (Mouse) 11438
Molecular Function: GABA receptor acetylcholine receptor ion channel ligand-gated ion channel receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.