Abnova
Manufacturer Code:H00001137B01P
Catalog # 89-002-542
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | CHRNA4 (AAH96292.1 1 a.a. ~ 451 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a full-length human CHRNA4 protein.
For Research Use Only
Protein Aliases: BFNC; cholinergic receptor, nicotinic alpha 4; cholinergic receptor, nicotinic, alpha 4 (neuronal); cholinergic receptor, nicotinic, alpha polypeptide 4; EBN; EBN1; FLJ95812; NACHR; NACHRA4; NACRA4; Neuronal acetylcholine receptor subunit alpha-4; neuronal nicotinic acetylcholine receptor alpha-4 subunit
Gene Aliases: BFNC; CHRNA4; EBN; EBN1; NACHR; NACHRA4; NACRA4
UniProt ID: (Human) P43681
Entrez Gene ID: (Human) 1137
Molecular Function: GABA receptor acetylcholine receptor ion channel ligand-gated ion channel receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.