Invitrogen
Manufacturer Code:PA577501
Catalog # PIPA577501
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide (C)KWKPSDYQGVEFMR corresponding to amino acid residues 91-104 of rat nAChRalpha3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acetylcholine receptor alpha 3 (neuronal nicotine); alpha 3; cetylcholine receptor alpha 3 (neuronal nicotine); cholinergic receptor nicotinic alpha polypeptide 4; cholinergic receptor, nicotinic, alpha polypeptide 3; nAChR alpha 3 CHRNA3; Neuronal acetylcholine receptor subunit alpha-3; neuronal nicotinic acetylcholine receptor, alpha 3 subunit
Gene Aliases: (a)3; A730007P14Rik; Acra-3; Acra3; Chrna3
UniProt ID: (Mouse) Q8R4G9
Entrez Gene ID: (Mouse) 110834, (Rat) 25101
Molecular Function:
GABA receptor
acetylcholine receptor
ion channel
ligand-gated ion channel
receptor
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.