Invitrogen
Manufacturer Code:PA5116458
Catalog # PIPA5116458
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide within rat CHRNA3 aa 80-120 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: (a)3 A730007P14Rik acetylcholine receptor Acetylcholine receptor alpha 3 (neuronal nicotine) Acra3 Acra-3 alpha 3 cetylcholine receptor alpha 3 (neuronal nicotine) cholinergic receptor cholinergic receptor nicotinic alpha 3 subunit cholinergic receptor nicotinic alpha polypeptide 4 cholinergic receptor nicotinic alpha 3 cholinergic receptor nicotinic alpha 3 (neuronal) cholinergic receptor nicotinic alpha polypeptide 3 Chrna3 LNCR2 nAChR alpha 3 NACHRA3 Neuronal acetylcholine receptor subunit alpha-3 neuronal nicotinic acetylcholine receptor alpha 3 subunit neuronal nicotinic acetylcholine receptor alpha3 subunit PAOD2; Acetylcholine receptor alpha 3 (neuronal nicotine); alpha 3; cetylcholine receptor alpha 3 (neuronal nicotine); cholinergic receptor nicotinic alpha polypeptide 4; cholinergic receptor, nicotinic alpha 3; cholinergic receptor, nicotinic, alpha 3 (neuronal); cholinergic receptor, nicotinic, alpha polypeptide 3; Neuronal acetylcholine receptor subunit alpha-3; neuronal nicotinic acetylcholine receptor, alpha 3 subunit; neuronal nicotinic acetylcholine receptor, alpha3 subunit
Gene Aliases: (a)3; A730007P14Rik; Acra-3; Acra3; CHRNA3; LNCR2; NACHRA3; PAOD2
UniProt ID: (Human) P32297, (Mouse) Q8R4G9
Entrez Gene ID: (Human) 1136, (Mouse) 110834, (Rat) 25101
Molecular Function: GABA receptor acetylcholine receptor ion channel ligand-gated ion channel receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.