Invitrogen
Manufacturer Code:PA584868
Catalog # PIPA584868
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human CHRNA1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acetylcholine receptor subunit alpha; acetylcholine receptor, nicotinic, alpha 1 (muscle); cholinergic receptor, nicotinic alpha 1; cholinergic receptor, nicotinic, alpha 1 (muscle); cholinergic receptor, nicotinic, alpha polypeptide 1 (muscle); CHRNA; muscle nicotinic acetylcholine receptor; nicotinic acetylcholine receptor alpha subunit; nicotinic cholinergic receptor alpha 1
Gene Aliases: ACHRA; ACHRD; CHNRA; CHRNA; CHRNA1; CMS1A; CMS1B; CMS2A; FCCMS; SCCMS
UniProt ID: (Human) P02708
Entrez Gene ID: (Human) 1134
Molecular Function: GABA receptor acetylcholine receptor ion channel ligand-gated ion channel receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.