Proteintech
Manufacturer Code:106131AP150UL
Catalog # 50-172-6448
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CHRNA1 Fusion Protein Ag0933 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acetylcholine receptor subunit alpha; acetylcholine receptor, nicotinic, alpha 1 (muscle); ACHRA ACHRD CHNRA CHRNA CHRNA1 CMS2A FCCMS nAChRA1 SCCMS; cholinergic receptor nicotinic alpha polypeptide 1; cholinergic receptor, nicotinic alpha 1; cholinergic receptor, nicotinic, alpha 1 (muscle); cholinergic receptor, nicotinic, alpha polypeptide 1 (muscle); muscle nicotinic acetylcholine receptor; nicotinic acetylcholine receptor alpha subunit; nicotinic cholinergic receptor alpha 1
Gene Aliases: Achr-1; ACHRA; ACHRD; Acra; AI385656; AI608266; CHNRA; CHRNA; CHRNA1; CMS1A; CMS1B; CMS2A; FCCMS; SCCMS
UniProt ID: (Human) P02708, (Mouse) P04756, (Rat) P25108
Entrez Gene ID: (Human) 1134, (Mouse) 11435, (Rat) 79557
Molecular Function:
GABA receptor
acetylcholine receptor
ion channel
ligand-gated ion channel
receptor
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.