Invitrogen
Manufacturer Code:PA577484
Catalog # PIPA577484
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide (C)HNATTVNGTPVNHQPLER corresponding to amino acid residues 7-24 of human M5 muscarinic receptor |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: acetylcholine receptor, muscarinic 5; cholinergic receptor, muscarinic 5; CHRM5; Muscarinic acetylcholine receptor M5
Gene Aliases: Chrm-5; CHRM5; HM5
UniProt ID: (Human) P08912, (Rat) P08911
Entrez Gene ID: (Human) 1133, (Rat) 53949
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.