Invitrogen
Manufacturer Code:PA577483
Catalog # PIPA577483
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide (C) RTLNPASKWSK corresponding to amino acid residues 330-340 of rat M4 Muscarinic Receptor |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: acetylcholine receptor, muscarinic 4; AChR M4; cholinergic receptor, muscarin 4; cholinergic receptor, muscarinic 4; CHRM4; Mm4 mAChR; muscarinic acetylcholine receptor 4; Muscarinic acetylcholine receptor M4
Gene Aliases: Chrm-4; CHRM4; HM4; M4; M4R
UniProt ID: (Human) P08173, (Mouse) P32211, (Rat) P08485
Entrez Gene ID: (Human) 1132, (Mouse) 12672, (Rat) 25111
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.