Invitrogen
Manufacturer Code:PA577485
Catalog # PIPA577485
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide (C)TLAKRFALKTRSQITKRKR corresponding to amino residues 461-479 of rat CHRM3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: acetylcholine receptor, muscarinic 3; AChR M3; cholinergic receptor, muscarinic 3; cholinergic receptor, muscarinic 3, cardiac; CHRM3; m3 muscarinic receptor; Mm3 mAChR; muscarinic acetylcholine receptor 3; Muscarinic acetylcholine receptor M3
Gene Aliases: Chrm-3; CHRM3; EGBRS; HM3; M3; M3R; PBS
UniProt ID: (Human) P20309, (Mouse) Q9ERZ3, (Rat) P08483
Entrez Gene ID: (Human) 1131, (Mouse) 12671, (Rat) 24260
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.