Invitrogen
Manufacturer Code:PA532708
Catalog # PIPA532708
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic 18 amino acid peptide from 3rd cytoplasmic domain of human CHRM3 / M3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: acetylcholine receptor, muscarinic 3; cholinergic receptor, muscarinic 3; EGBRS HM3 acetylcholine receptor muscarinic 3 m3 muscarinic receptor muscarinic acetylcholine receptor M3; m3 muscarinic receptor; Muscarinic acetylcholine receptor M3
Gene Aliases: CHRM3; EGBRS; HM3; PBS
UniProt ID: (Human) P20309
Entrez Gene ID: (Human) 1131
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.