Invitrogen
Manufacturer Code:PA186940
Catalog # PIPA186940
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide: 3rd cytoplasmic domain of human muscarinic receptor M3 conjugated to KLH. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4°C or -20°C if preferred |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: acetylcholine receptor, muscarinic 3; cholinergic receptor, muscarinic 3; HM3; m3 muscarinic receptor; Muscarinic acetylcholine receptor M3
Gene Aliases: CHRM3; EGBRS; HM3; PBS
UniProt ID: (Human) P20309
Entrez Gene ID: (Human) 1131
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.