Abnova
Manufacturer Code:PAB16211
Catalog # 89-118-101
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide (conjugated with KLH) corresponding to human CHRM3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of CHRM3.
For Research Use Only
Protein Aliases: acetylcholine receptor, muscarinic 3; cholinergic receptor, muscarinic 3; HM3; m3 muscarinic receptor; Muscarinic acetylcholine receptor M3
Gene Aliases: CHRM3; EGBRS; HM3; PBS
UniProt ID: (Human) P20309
Entrez Gene ID: (Human) 1131
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.