Invitrogen
Manufacturer Code:MA535662
Catalog # PIMA535662
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC1225 |
Immunogen | A synthesized peptide derived from human ACM2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: 7TM receptor; acetylcholine receptor, muscarinic 2; AChR M2; AChR-M2 Acm2 CHRM2 Chrm-2 FLJ43243 HM2 M2 m2AChR M2-mAChR MGC120006 MGC120007 Muscarinic acetylcholine receptor M2; cholinergic receptor, muscarinic 2; cholinergic receptor, muscarinic 2, cardiac; cholinergic receptor, muscarinic 2, isoform a; M2 muscarinic acetylcholine receptor; muscarinic acetylcholine receptor 2; Muscarinic acetylcholine receptor M2; muscarinic M2 receptor; muscarinic receptor m2
Gene Aliases: AChR-M2; Acm2; Chrm-2; CHRM2; HM2; M2
UniProt ID: (Human) P08172, (Mouse) Q9ERZ4, (Rat) P10980
Entrez Gene ID: (Human) 1129, (Mouse) 243764, (Rat) 81645
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.