Invitrogen
Manufacturer Code:PA533430
Catalog # PIPA533430
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic 20 amino acid peptide from 3rd cytoplasmic domain of human CHRM2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 7TM receptor; acetylcholine receptor, muscarinic 2; cholinergic receptor, muscarinic 2; cholinergic receptor, muscarinic 2, isoform a; CHRM2 7TM receptor HM2 M2 muscarinic receptor M2-mAChR Muscarinic receptor M2 Muscarinic M2 receptor; Muscarinic acetylcholine receptor M2; muscarinic M2 receptor
Gene Aliases: CHRM2; HM2
UniProt ID: (Human) P08172
Entrez Gene ID: (Human) 1129
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.