Invitrogen
Manufacturer Code:PA5111845
Catalog # PIPA5111845
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | GST fusion protein corresponding to amino acid residues 225-356 of human M2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 7TM receptor; acetylcholine receptor, muscarinic 2; AChR M2; AChR-M2 Acm2 CHRM2 Chrm-2 FLJ43243 HM2 M2 m2AChR M2-mAChR MGC120006 MGC120007 Muscarinic acetylcholine receptor M2; cholinergic receptor, muscarinic 2; cholinergic receptor, muscarinic 2, cardiac; cholinergic receptor, muscarinic 2, isoform a; M2 muscarinic acetylcholine receptor; muscarinic acetylcholine receptor 2; Muscarinic acetylcholine receptor M2; muscarinic M2 receptor; muscarinic receptor m2
Gene Aliases: AChR-M2; Acm2; Chrm-2; CHRM2; HM2; M2
UniProt ID: (Human) P08172, (Mouse) Q9ERZ4, (Rat) P10980
Entrez Gene ID: (Human) 1129, (Mouse) 243764, (Rat) 81645
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.