Invitrogen
Manufacturer Code:PA577482
Catalog # PIPA577482
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | GST fusion protein with a sequence GSETPGKGGGSSSSSERSQP GAEGSPETPPGRCCRCCRAPRLLQAYSWKEEEEEDEGSMESLTSS EGEEPGSEVVIKMPMVDPEAQAPTKQPPRSSPNTVKRPTKKGRDR AGKGQKPRGKEQLAKRK corresponding to amino acid residues 227-353 of human m1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: acetylcholine receptor, muscarinic 1; AChR M1; Cholinergic receptor, muscarin 1; cholinergic receptor, muscarinic 1; CHRM1; m1 muscarinic acetylcholine receptor protein; Muscarinic acetylcholine receptor M1
Gene Aliases: Chrm-1; CHRM1; HM1; M1; M1R
UniProt ID: (Human) P11229, (Mouse) P12657, (Rat) P08482
Entrez Gene ID: (Human) 1128, (Mouse) 12669, (Rat) 25229
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.