Novus Biologicals
Manufacturer Code:NBP237304
Catalog # NBP237304
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 5D12D1 |
Immunogen | Purified recombinant fragment of human MLXIPL (AA: 18-143) expressed in E. Coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Protein Aliases: bHLHd14; BHLHD14 CHREBPcarbohydrate-responsive element-binding protein Class D basic helix-loop-helix protein 14 MIOWS basic-helix-loop-helix leucine zipper protein MLX interacting protein-like MLX interactor MLX-interacting protein-like MONDOBWilliams-Beuren syndrome chromosomal region 14 protein WBSCR14Williams-Beuren syndrome chromosome region 14 protein 1 Williams Beuren syndrome chromosome region 14 Williams-Beuren syndrome chromosome region 14 protein 2 WS-bHLHbHLHd14carbohydrate response element binding protein; carbohydrate response element binding protein; Carbohydrate-responsive element-binding protein; ChREBP; Class D basic helix-loop-helix protein 14; MLX interacting protein-like; MLX interactor; MLX-interacting protein-like; Williams Beuren syndrome chromosome region 14; Williams-Beuren syndrome chromosomal region 14 protein; Williams-Beuren syndrome chromosome region 14 protein 1; Williams-Beuren syndrome chromosome region 14 protein 2; Williams-Beuren syndrome chromosome region 14 protein 3; WS basic-helix-loop-helix leucine zipper protein; WS-bHLH
Gene Aliases: BHLHD14; CHREBP; MIO; MLXIPL; MONDOB; WBSCR14; WS-bHLH
UniProt ID: (Human) Q9BY05
Entrez Gene ID: (Human) 51085
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.