Invitrogen
Manufacturer Code:PA536445
Catalog # PIPA536445
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 640-690 of human CHSY2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Chondroitin glucuronyltransferase 2; Chondroitin sulfate synthase 2; Chondroitin sulfate synthase 2 Chondroitin glucuronyltransferase 2 Chondroitin-polymerizing factor ChPF Glucuronosyl-N-acetylgalactosaminyl-proteoglycan 4-beta-N-acetylgalactosaminyltransferase II N-acetylgalactosaminyl-proteoglycan 3-beta-glucuronosyltransferase II N-acetylgalactosaminyltransferase 2 CHPF CSS2 ORF Names: UNQ651/PRO1281; Chondroitin-polymerizing factor; ChPF; Glucuronosyl-N-acetylgalactosaminyl-proteoglycan 4-beta-N-acetylgalactosaminyltransferase II; N-acetylgalactosaminyl-proteoglycan 3-beta-glucuronosyltransferase II; N-acetylgalactosaminyltransferase 2
Gene Aliases: 1700028N03Rik; AI414328; CHPF; CHSY2; CSS2; D1Bwg1363e; UNQ651/PRO1281
UniProt ID: (Human) Q8IZ52, (Mouse) Q6IQX7
Entrez Gene ID: (Human) 79586, (Mouse) 74241, (Rat) 316533
Molecular Function: glycosyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.