OriGene
Manufacturer Code:TA810172
Catalog # 50-168-3237
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1D11 |
Immunogen | Full length human recombit protein of human CHP1 produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: calcineurin B homolog; Calcineurin B homologous protein 1; Calcineurin B-like protein; calcineurin homologous protein; calcineurin-like EF hand protein 1; calcineurin-like EF-hand protein 1; calcium binding protein P22; Calcium-binding protein CHP; Calcium-binding protein p22; CHP SLC9A1BP Sid470p p22 p24; EF-hand calcium-binding domain-containing protein p22; SLC9A1 binding protein
Gene Aliases: CHP; CHP1; p22; p24; Sid470p; SLC9A1BP
UniProt ID: (Human) Q99653
Entrez Gene ID: (Human) 11261
Molecular Function: calcium-binding protein calmodulin hydrolase intracellular calcium-sensing protein phosphatase protein phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.