Invitrogen
Manufacturer Code:PA543799
Catalog # PIPA543799
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human CHP |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: calcineurin B homolog; Calcineurin B homologous protein 1; Calcineurin B-like protein; calcineurin homologous protein; calcineurin-like EF hand protein 1; calcineurin-like EF-hand protein 1; calcium binding protein P22; Calcium binding protein P22 SLC9A1BP CHP; Calcium-binding protein CHP; Calcium-binding protein p22; EF-hand Ca2+-binding protein p22; EF-hand calcium-binding domain-containing protein p22; p24; Sid 470; SLC9A1 binding protein
Gene Aliases: 1500003O03Rik; AA960066; AI046351; Cahp; CHP; CHP1; p22; p24; si:dkey-41n20.1; Sid470; Sid470p; SLC9A1BP; vac; Wrch2; wu:fc69b04; zgc:63904
UniProt ID: (Bovine) Q3SYS6, (Human) Q99653, (Rat) P61023, (Mouse) P61022
Entrez Gene ID: (Dog) 478264, (Rabbit) 100344087, (Bovine) 512662, (Human) 11261, (Horse) 100056957, (Guinea pig) 100719349, (Rat) 64152, (Mouse) 56398, (Zebrafish) 325361
Molecular Function:
calcium-binding protein
calmodulin
hydrolase
intracellular calcium-sensing protein
phosphatase
protein phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.