Invitrogen
Manufacturer Code:PA5106622
Catalog # PIPA5106622
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human CHP corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1500003O03Rik AA960066 AI046351 Cahp calcineurin B homolog calcineurin B homologous protein 1 Calcineurin B-like protein calcineurin homologous protein calcineurin like EF-hand protein 1 calcineurin-like EF hand protein 1 calcineurin-like EF-hand protein 1 calcium binding protein P22 calcium-binding protein CHP Calcium-binding protein p22 CHP CHP1 Chromo domain-containing protein 1 chromodomain protein Chp1 EF-hand Ca2+-binding protein p22 EF-hand calcium-binding domain-containing protein p22 p22 p24 sid 470 Sid470 Sid470p SLC9A1 binding protein SLC9A1BP SPAC18G6.02c vac Wrch2; calcineurin B homolog; Calcineurin B homologous protein 1; Calcineurin B-like protein; calcineurin homologous protein; calcineurin-like EF hand protein 1; calcineurin-like EF-hand protein 1; calcium binding protein P22; Calcium-binding protein CHP; Calcium-binding protein p22; EF-hand Ca2+-binding protein p22; EF-hand calcium-binding domain-containing protein p22; p24; Sid 470; SLC9A1 binding protein
Gene Aliases: 1500003O03Rik; AA960066; AI046351; Cahp; CHP; CHP1; p22; p24; Sid470; Sid470p; SLC9A1BP; vac; Wrch2
UniProt ID: (Human) Q99653, (Rat) P61023, (Mouse) P61022
Entrez Gene ID: (Human) 11261, (Rat) 64152, (Mouse) 56398
Molecular Function:
calcium-binding protein
calmodulin
hydrolase
intracellular calcium-sensing protein
phosphatase
protein phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.