Invitrogen
Manufacturer Code:MA521318
Catalog # PIMA521318
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3G8 |
Immunogen | CHORDC1 (AAH08373 1 a.a. &bsim 66 a.a) full-length recombit protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CHORD domain-containing protein 1; CHORD-containing protein 1; CHP-1; CHP1 CHORD-containing protein 1 CHP-1 chord domain-containing protein 1 cysteine and histidine-rich domain (CHORD)-containing 1 cysteine and histidine-rich domain (CHORD)-containing zinc-binding protein 1 cysteine and histidine-rich domain-containing protein 1 protein morgana; cysteine and histidine-rich domain (CHORD) containing 1; cysteine and histidine-rich domain (CHORD)-containing 1; cysteine and histidine-rich domain (CHORD)-containing, zinc-binding protein 1; Cysteine and histidine-rich domain-containing protein 1; morgana; Protein morgana
Gene Aliases: CHORDC1; CHP1
UniProt ID: (Human) Q9UHD1
Entrez Gene ID: (Human) 26973
Molecular Function:
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.