Invitrogen
Manufacturer Code:PA544222
Catalog # PIPA544222
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human CHORDC1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CHORD domain-containing protein 1; CHORD-containing protein 1; CHP-1; cysteine and histidine-rich domain (CHORD) containing 1; Cysteine and histidine-rich domain (CHORD) containing 1 CHP1; cysteine and histidine-rich domain (CHORD)-containing 1; cysteine and histidine-rich domain (CHORD)-containing, zinc binding protein 1; cysteine and histidine-rich domain (CHORD)-containing, zinc-binding protein 1; Cysteine and histidine-rich domain-containing protein 1; Morgana; Protein morgana
Gene Aliases: 1110001O09Rik; AA409036; CHORDC1; CHP-1; CHP1; Morgana; zgc:63894
UniProt ID: (Bovine) Q29RL2, (Human) Q9UHD1, (Rat) D4A4T9, (Mouse) Q9D1P4, (Zebrafish) Q7T3F7
Entrez Gene ID: (Bovine) 505144, (Human) 26973, (Rabbit) 100339204, (Dog) 476773, (Horse) 100050120, (Rat) 315447, (Mouse) 66917, (Guinea pig) 100726003, (Zebrafish) 393310
Molecular Function:
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.