Novus Biologicals
Manufacturer Code:NBP178304SS
Catalog # NBP178304SS
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide made to an internal portion of the human CHORDC1 protein (between residues 100-250) [UniProt Q9UHD1] |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CHORD domain-containing protein 1; CHORD domain-containing protein 1 CHORD-containing protein 1 CHP1CHP-1 cysteine and histidine-rich domain (CHORD) containing 1 cysteine and histidine-rich domain (CHORD)-containing 1 cysteine and histidine-rich domain (CHORD)-containing zinc-binding protein 1 cysteine and histidine-rich domain-containing protein 1 FLJ37289 Protein morgana; CHORD-containing protein 1; CHP-1; cysteine and histidine-rich domain (CHORD) containing 1; cysteine and histidine-rich domain (CHORD)-containing 1; cysteine and histidine-rich domain (CHORD)-containing, zinc-binding protein 1; Cysteine and histidine-rich domain-containing protein 1; morgana; Protein morgana
Gene Aliases: CHORDC1; CHP1
UniProt ID: (Human) Q9UHD1
Entrez Gene ID: (Human) 26973
Molecular Function:
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.