OriGene
Manufacturer Code:TA505096
Catalog # 50-167-5643
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2A1 |
Immunogen | Full length human recombit protein of human CHN1 produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: A-chimaerin; a2-chimaerin; Alpha-chimerin; ARHGAP2 CHN DURS2 NC RHOGAP2; chimaerin 1; chimerin (chimaerin) 1; N-chimaerin; N-chimerin; NC; Rho GTPase-activating protein 2
Gene Aliases: ARHGAP2; CHN; CHN1; DURS2; NC; RHOGAP2
UniProt ID: (Human) P15882
Entrez Gene ID: (Human) 1123
Molecular Function:
G-protein modulator
enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.